Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | XM_011543916.2 | 2 | 254 | XP_011542218.1 |
XM_017029565.1 | 2 | 254 | XP_016885054.1 | |
XM_017029566.1 | 2 | 254 | XP_016885055.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | PPP1R3F CCDC22 CACNA1F FOXP3 GAGE10 |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | PRICKLE3 HDAC6 LOC101927635 SUV39H1 TFE3 ERAS CCDC22 GPKOW OTUD5 WAS GATA1 SYP PLP2 FOXP3 PIM2 MAGIX GRIPAP1 SLC35A2 CCDC120 WDR45 TBC1D25 CACNA1F PCSK1N EBP RBM3 PQBP1 TIMM17B PPP1R3F SYP-AS1 GLOD5 PORCN PRAF2 SLC38A5 FTSJ1 WDR13 KCND1 |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX9 SSX4B PRICKLE3 ZNF630-AS1 HDAC6 LOC101927635 SPACA5B SUV39H1 TFE3 ERAS CCDC22 GPKOW SSX5 OTUD5 WAS GATA1 SYP PLP2 FOXP3 PIM2 MAGIX SPACA5 GRIPAP1 ZNF182 SLC35A2 CCDC120 WDR45 TBC1D25 CACNA1F PCSK1N EBP RBM3 PQBP1 TIMM17B PPP1R3F SYP-AS1 GLOD5 PORCN SSX4 PRAF2 SSX6 SSX3 SLC38A5 FTSJ1 WDR13 KCND1 ZNF630 SSX1 |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | PPP1R3F CCDC22 CACNA1F FOXP3 GAGE10 |