Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OPRL1
Entrez Gene ID
4987
Gene Name
opioid related nociceptin receptor 1
Gene Aliases
KOR-3, NOCIR, NOP, NOPr, OOR, ORL1
Chromosome Location
Chr.20: 64080082 - 64100643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001200019.1 NP_001186948.1 3 - 4 375 59
NM_001318853.1 NP_001305782.1 2 - 3 418 59
NM_182647.3 NP_872588.1 2 - 3 418 59
XM_017027855.1 XP_016883344.1 2 - 3 399 59
GenBank mRNA AF285095.1 - 2 - 3 327 59
BC038433.1 - 3 - 4 375 59
U30185.1 - 1 - 2 80 59
X77130.1 - 1 - 2 140 59

View Full Product Details