The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200030.1 | NP_001186959.1 | 1 - 2 | 253 | 65 |
NM_003549.3 | NP_003540.2 | 1 - 2 | 253 | 65 | |
GenBank mRNA | AF036035.1 | - | 1 - 2 | 253 | 65 |
AF040710.1 | - | 1 - 2 | 173 | 65 | |
AK313455.1 | - | 1 - 2 | 164 | 65 |