The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013270.4 | NP_037402.1 | 2 - 3 | 367 | 87 |
GenBank mRNA | AF100707.1 | - | 2 - 3 | 367 | 87 |
BC033016.1 | - | 2 - 3 | 358 | 87 | |
BC037775.2 | - | 2 - 3 | 329 | 87 |