The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024674.5 | NP_078950.1 | 2 - 3 | 356 | 76 |
XM_011542148.1 | XP_011540450.1 | 2 - 3 | 346 | 76 | |
GenBank mRNA | AF521099.1 | - | 2 - 3 | 307 | 76 |
AK022519.1 | - | 2 - 3 | 307 | 76 | |
BC028566.2 | - | 2 - 3 | 346 | 76 | |
CN349325.1 | - | 2 - 3 | 358 | 76 |