Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAGE5
Entrez Gene ID
2577
Gene Name
G antigen 5
Gene Aliases
CT4.5
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001475.1 NP_001466.1 3 - 4 281 91
GenBank mRNA BC024914.1 - 3 - 4 294 91
U19146.1 - 3 - 4 281 91

Gene Symbol
GAGE1
Entrez Gene ID
2543
Gene Name
G antigen 1
Gene Aliases
CT4.1, GAGE-1
Chromosome Location
Chr.X: 49598992 - 49608536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040663.2 NP_001035753.1 3 - 4 330 91
NR_102272.1 3 - 4 330 91
XM_011543896.2 XP_011542198.1 3 - 4 351 91
GenBank mRNA BC005363.1 - 3 - 4 324 91
BC036094.1 - 3 - 4 318 91
BC069470.1 - 3 - 4 281 91
DB448801.1 - 3 - 4 294 91
DC401704.1 - 3 - 4 330 91
U19142.1 - 3 - 4 252 91

Gene Symbol
GAGE12B
Entrez Gene ID
729428
Gene Name
G antigen 12B
Gene Aliases
GAGE-12B
Chromosome Location
Chr.X: 49341183 - 49529921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127345.1 NP_001120817.1 3 - 4 288 91
GenBank mRNA BC130374.1 - 3 - 4 288 91
BC143615.1 - 3 - 4 288 91
BC143619.1 - 3 - 4 288 91

Gene Symbol
GAGE12C
Entrez Gene ID
729422
Gene Name
G antigen 12C
Gene Aliases
GAGE-12B
Chromosome Location
Chr.X: 49532177 - 49539541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098408.1 NP_001091878.1 3 - 4 323 91

Gene Symbol
GAGE12D
Entrez Gene ID
100132399
Gene Name
G antigen 12D
Gene Aliases
GAGE-12B
Chromosome Location
Chr.X: 49541722 - 49549107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127199.1 NP_001120671.1 3 - 4 334 91

Gene Symbol
GAGE12E
Entrez Gene ID
729431
Gene Name
G antigen 12E
Gene Aliases
GAGE-12B
Chromosome Location
Chr.X: 49551289 - 49558649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098418.1 NP_001091888.1 3 - 4 323 91

Gene Symbol
GAGE12F
Entrez Gene ID
100008586
Gene Name
G antigen 12F
Gene Aliases
-
Chromosome Location
Chr.X: 49560808 - 49568208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098405.2 NP_001091875.1 3 - 4 357 91
GenBank mRNA AI381509.1 - 2 - 3 91
CF780547.1 - 3 - 4 280 91
HY016295.1 - 3 - 4 357 91

Gene Symbol
GAGE12G
Entrez Gene ID
645073
Gene Name
G antigen 12G
Gene Aliases
AL4, CT4.7, GAGE-12G
Chromosome Location
Chr.X: 49570400 - 49577757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098409.1 NP_001091879.1 3 - 4 323 91

Gene Symbol
GAGE12I
Entrez Gene ID
26748
Gene Name
G antigen 12I
Gene Aliases
AL4, CT4.7, GAGE-7B, GAGE7B
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001477.1 NP_001468.1 3 - 4 261 91

Gene Symbol
GAGE12J
Entrez Gene ID
729396
Gene Name
G antigen 12J
Gene Aliases
GAGE11
Chromosome Location
Chr.X: 49322030 - 49329387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098406.2 NP_001091876.2 3 - 4 330 91
GenBank mRNA AK310423.1 - 3 - 4 330 91
BX108227.1 - 3 - 4 271 91

Gene Symbol
GAGE13
Entrez Gene ID
645051
Gene Name
G antigen 13
Gene Aliases
GAGE-12A, GAGE-13, GAGE12A
Chromosome Location
Chr.X: 49331603 - 49338952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098412.2 NP_001091882.2 3 - 4 302 91

Gene Symbol
GAGE2A
Entrez Gene ID
729447
Gene Name
G antigen 2A
Gene Aliases
CT4.2, GAGE-2, GAGE-2A, GAGE2
Chromosome Location
Chr.X: 49589515 - 49596827 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC069309.1 - 3 - 4 276 91
BC069558.1 - 3 - 4 254 91

Gene Symbol
GAGE2B
Entrez Gene ID
645037
Gene Name
G antigen 2B
Gene Aliases
CT4.2, GAGE-2, GAGE-2B
Chromosome Location
Chr.X: 49331615 - 49338933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098411.3 NP_001091881.1 3 - 4 287 91

Gene Symbol
GAGE2C
Entrez Gene ID
2574
Gene Name
G antigen 2C
Gene Aliases
CT4.2, GAGE-2, GAGE-2C, GAGE2
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001472.2 NP_001463.1 3 - 4 287 91
GenBank mRNA BC113926.1 - 3 - 4 258 91
U19143.1 - 3 - 4 287 91

Gene Symbol
GAGE2D
Entrez Gene ID
729408
Gene Name
G antigen 2D
Gene Aliases
CT4.8, GAGE-2D, GAGE-8, GAGE8
Chromosome Location
Chr.X: 49560844 - 49568208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098407.1 NP_001091877.1 3 - 4 320 91

Gene Symbol
GAGE2E
Entrez Gene ID
26749
Gene Name
G antigen 2E
Gene Aliases
GAGE-2E, GAGE8
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127200.2 NP_001120672.2 3 - 4 320 91
GenBank mRNA BC018052.1 - 3 - 4 300 91
HY204089.1 - 2 - 3 91

Gene Symbol
GAGE3
Entrez Gene ID
2575
Gene Name
G antigen 3
Gene Aliases
CT4.3
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA U19144.1 - - 309 91

Gene Symbol
GAGE4
Entrez Gene ID
2576
Gene Name
G antigen 4
Gene Aliases
CT4.4
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001474.1 NP_001465.1 3 - 4 289 91
GenBank mRNA BC171769.1 - 3 - 4 255 91
CR542236.1 - 2 - 3 207 91
U19145.1 - 3 - 4 289 91

Gene Symbol
GAGE6
Entrez Gene ID
2578
Gene Name
G antigen 6
Gene Aliases
CT4.6
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001476.1 NP_001467.1 3 - 4 288 91
GenBank mRNA U19147.1 - 3 - 4 288 91

Gene Symbol
GAGE7
Entrez Gene ID
2579
Gene Name
G antigen 7
Gene Aliases
AL4, CT4.7, GAGE-7
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021123.2 NP_066946.1 3 - 4 260 91
GenBank mRNA AF055474.1 - 3 - 4 261 91
AF058988.1 - 3 - 4 286 91
BC031628.1 - 3 - 4 279 91
BC104951.1 - 3 - 4 288 91
BC121182.1 - 3 - 4 267 91
BC121183.1 - 3 - 4 267 91
BC144193.1 - 3 - 4 255 91

Gene Symbol
GAGE8
Entrez Gene ID
100101629
Gene Name
G antigen 8
Gene Aliases
CT4.8, GAGE-8
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012196.1 NP_036328.1 3 - 4 276 91
GenBank mRNA AF055473.1 - 3 - 4 276 91
BC143620.1 - 3 - 4 285 91

View Full Product Details