Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- POTEH
- Entrez Gene ID
-
23784
- Gene Name
- POTE ankyrin domain family member H
- Gene Aliases
-
A26C3, ACTBL1, CT104.7, POTE22
- Chromosome Location
-
Chr.22: 15690026 - 15721631 on Build GRCh38
- Gene Symbol
- POTEG
- Entrez Gene ID
-
404785
- Gene Name
- POTE ankyrin domain family member G
- Gene Aliases
-
A26C2, ACTBL1, CT104.4, POTE-14, POTE14, POTE14alpha, POTE22
- Chromosome Location
-
Chr.14: 19402764 - 19434341 on Build GRCh38
- Gene Symbol
- POTEM
- Entrez Gene ID
-
641455
- Gene Name
- POTE ankyrin domain family member M
- Gene Aliases
-
ACT, P704P, POTE14beta
- Chromosome Location
-
Chr.14: 18967434 - 19003752 on Build GRCh38
View Full Product Details