Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NBPF11
- Entrez Gene ID
-
200030
- Gene Name
- neuroblastoma breakpoint family member 11
- Gene Aliases
-
NBPF24
- Chromosome Location
-
Chr.1: 148102046 - 148152190 on Build GRCh38
- Gene Symbol
- NBPF10
- Entrez Gene ID
-
100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
-
AB6, AG1, NBPF9
- Chromosome Location
-
Chr.1: 146066318 - 146144804 on Build GRCh38
- Gene Symbol
- NBPF12
- Entrez Gene ID
-
149013
- Gene Name
- neuroblastoma breakpoint family member 12
- Gene Aliases
-
COAS1, KIAA1245
- Chromosome Location
-
Chr.1: 146938545 - 146996093 on Build GRCh38
- Gene Symbol
- NBPF14
- Entrez Gene ID
-
25832
- Gene Name
- neuroblastoma breakpoint family member 14
- Gene Aliases
-
DJ328E19.C1.1, NBPF
- Chromosome Location
-
Chr.1: 148531385 - 148607636 on Build GRCh38
- Gene Symbol
- NBPF19
- Entrez Gene ID
-
101060226
- Gene Name
- neuroblastoma breakpoint family member 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 149472955 - 149556256 on Build GRCh38
- Gene Symbol
- NBPF8
- Entrez Gene ID
-
728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
-
NBPF8P
- Chromosome Location
-
Chr.1: 120436351 - 120467844 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY894568.1 |
- |
7 - 8 |
1063 |
112 |
AY894569.1 |
- |
7 - 8 |
1065 |
112 |
AY894571.1 |
- |
6 - 7 |
1165 |
112 |
AY894573.1 |
- |
6 - 7 |
1165 |
112 |
- Gene Symbol
- NBPF9
- Entrez Gene ID
-
400818
- Gene Name
- neuroblastoma breakpoint family member 9
- Gene Aliases
-
AE01
- Chromosome Location
-
Chr.1: 149054033 - 149090861 on Build GRCh38
View Full Product Details