Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCE2B
Entrez Gene ID
26239
Gene Name
late cornified envelope 2B
Gene Aliases
LEP10, SPRL1B, XP5
Chromosome Location
Chr.1: 152686123 - 152687401 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014357.4 NP_055172.1 2 - 2 551 93
GenBank mRNA AF086289.1 - 2 - 2 93

View Full Product Details