The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037161.1 | NP_001032238.1 | 1 - 1 | 148 | 87 |
GenBank mRNA | AK300886.1 | - | 1 - 1 | 185 | 87 |
BX161396.1 | - | 1 - 1 | 258 | 87 | |
DQ082754.1 | - | 1 - 1 | 148 | 87 |