The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001818.3 | NP_001809.3 | 7 - 8 | 884 | 72 |
GenBank mRNA | AB031085.1 | - | 7 - 8 | 851 | 72 |
AB045829.1 | - | 7 - 8 | 851 | 72 | |
AK314988.1 | - | 7 - 8 | 925 | 72 | |
BC020744.1 | - | 7 - 8 | 884 | 72 | |
D26125.1 | - | 7 - 8 | 847 | 72 | |
M33375.1 | - | 7 - 8 | 849 | 72 | |
S68287.1 | - | 7 - 8 | 858 | 72 |