The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004617.3 | NP_004608.1 | 4 - 5 | 885 | 63 |
GenBank mRNA | AK055577.1 | - | 4 - 5 | 1492 | 63 |
BC001386.2 | - | 4 - 5 | 755 | 63 | |
U31449.1 | - | 4 - 5 | 753 | 63 |