The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021724.4 | NP_068370.1 | 5 - 6 | 1876 | 86 |
GenBank mRNA | AK300155.1 | - | 2 - 3 | 572 | 86 |
AK315270.1 | - | 6 - 7 | 1618 | 86 | |
BC047875.1 | - | 5 - 6 | 1700 | 86 | |
BC056148.1 | - | 5 - 6 | 1767 | 86 | |
HQ692861.1 | - | 5 - 6 | 1245 | 86 | |
M24898.1 | - | 5 - 6 | 1872 | 86 | |
X72631.1 | - | 5 - 6 | 1447 | 86 |