The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145873.1 | NP_001139345.1 | 5 - 5 | 1266 | 65 |
NM_001768.6 | NP_001759.3 | 2 - 2 | 1124 | 65 | |
NM_171827.3 | NP_741969.1 | 2 - 2 | 1124 | 65 | |
NR_027353.1 | 3 - 3 | 710 | 65 | ||
GenBank mRNA | AK124156.1 | - | 5 - 5 | 1266 | 65 |
AK300089.1 | - | 3 - 3 | 676 | 65 | |
AY039664.1 | - | 2 - 2 | 414 | 65 | |
BC025715.1 | - | 2 - 2 | 240 | 65 | |
M12824.1 | - | 2 - 2 | 322 | 65 | |
M12828.1 | - | 2 - 2 | 300 | 65 |