Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA1
Entrez Gene ID
59338
Gene Name
pleckstrin homology domain containing A1
Gene Aliases
TAPP1
Chromosome Location
Chr.10: 122374578 - 122432355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195608.1 NP_001182537.1 12 - 13 1111 101
XM_005270019.1 XP_005270076.1 12 - 13 1170 101
XM_005270020.1 XP_005270077.1 12 - 13 1170 101
XM_005270021.4 XP_005270078.1 12 - 13 1170 101
XM_017016483.1 XP_016871972.1 12 - 13 1407 101
XM_017016484.1 XP_016871973.1 12 - 13 1348 101
XM_017016485.1 XP_016871974.1 11 - 12 1054 101
XM_017016486.1 XP_016871975.1 12 - 13 1348 101
XM_017016487.1 XP_016871976.1 12 - 13 1407 101
XM_017016488.1 XP_016871977.1 11 - 12 1054 101
XM_017016490.1 XP_016871979.1 12 - 13 1407 101
XM_017016491.1 XP_016871980.1 11 - 12 1054 101
GenBank mRNA AK057463.1 - 12 - 13 1111 101

View Full Product Details