Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC100129345
- Entrez Gene ID
-
100129345
- Gene Name
- uncharacterized LOC100129345
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 97632647 - 97686658 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033943.1 |
|
3 - 4 |
626 |
114 |
GenBank mRNA |
BX161483.1 |
- |
3 - 4 |
626 |
114 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX338081.2 |
- |
3 - 4 |
586 |
|
Chr. 14 - 97633228 - 97686619 [-] on Build GRCh38 |
114 |
GenBankmRNA |
BX361585.2 |
- |
3 - 4 |
626 |
|
Chr. 14 - 97633348 - 97686658 [-] on Build GRCh38 |
114 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101929241 |
LOC101929241 |
|
-51.3kb
|
Anti-sense |
Chr. - 97458816 - 97581383 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1550 |
LINC01550 |
C14orf64 |
279.2kb
|
Sense |
Chr. - 97925610 - 97978124 on Build GRCh38 |
View Full Product Details