The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_036433.2 | 1 - 2 | 283 | 67 | |
GenBank mRNA | AK127584.1 | - | 1 - 2 | 284 | 67 |
EG328077.1 | - | 1 - 2 | 67 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SUMO1/sentrin specific peptidase 7 | SENP7 | -5.0kb | Anti-sense | Chr. - 101324189 - 101513241 on Build GRCh38 | |
Downstream | tRNA methyltransferase 10C, mitochondrial RNase P subunit | TRMT10C | COXPD30, HNYA, MRPP1, RG9MTD1 | 37.9kb | Sense | Chr. - 101561836 - 101566446 on Build GRCh38 |