The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 65 - 66 | 11276 | 84 |
XM_006721206.2 | XP_006721269.1 | 65 - 66 | 11170 | 84 | |
XM_011523146.2 | XP_011521448.1 | 66 - 67 | 11305 | 84 | |
XM_011523147.1 | XP_011521449.1 | 66 - 67 | 11272 | 84 | |
XM_011523148.1 | XP_011521450.1 | 66 - 67 | 11345 | 84 | |
XM_011523151.1 | XP_011521453.1 | 65 - 66 | 11202 | 84 | |
XM_011523152.1 | XP_011521454.1 | 28 - 29 | 5029 | 84 | |
XM_011523155.2 | XP_011521457.1 | 21 - 22 | 4030 | 84 | |
XM_017023346.1 | XP_016878835.1 | 65 - 66 | 11448 | 84 | |
XM_017023347.1 | XP_016878836.1 | 53 - 54 | 9687 | 84 | |
XM_017023348.1 | XP_016878837.1 | 53 - 54 | 9427 | 84 |