The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 81 - 82 | 14298 | 72 |
XM_006721206.2 | XP_006721269.1 | 81 - 82 | 14192 | 72 | |
XM_011523146.2 | XP_011521448.1 | 82 - 83 | 14327 | 72 | |
XM_011523147.1 | XP_011521449.1 | 82 - 83 | 14294 | 72 | |
XM_011523148.1 | XP_011521450.1 | 82 - 83 | 14367 | 72 | |
XM_011523151.1 | XP_011521453.1 | 81 - 82 | 14224 | 72 | |
XM_011523152.1 | XP_011521454.1 | 44 - 45 | 8051 | 72 | |
XM_011523155.2 | XP_011521457.1 | 37 - 38 | 7052 | 72 | |
XM_017023346.1 | XP_016878835.1 | 81 - 82 | 14470 | 72 | |
XM_017023347.1 | XP_016878836.1 | 69 - 70 | 12709 | 72 | |
XM_017023348.1 | XP_016878837.1 | 69 - 70 | 12449 | 72 | |
GenBank mRNA | AK026688.1 | - | 2 - 3 | 393 | 72 |
AK074472.1 | - | 6 - 7 | 1117 | 72 | |
AL137259.1 | - | 9 - 10 | 1694 | 72 | |
BC043273.2 | - | 16 - 17 | 3018 | 72 |