Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR2
Entrez Gene ID
57047
Gene Name
phospholipid scramblase 2
Gene Aliases
-
Chromosome Location
Chr.3: 146391363 - 146497163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199978.1 NP_001186907.1 5 - 6 765 140
NM_001199979.1 NP_001186908.1 3 - 4 382 140
NM_020359.2 NP_065092.1 4 - 5 495 140
XM_011513013.2 XP_011511315.2 3 - 4 1545 140
XM_011513019.2 XP_011511321.2 3 - 4 1545 140
XM_011513020.2 XP_011511322.2 3 - 4 1545 140
XM_011513021.2 XP_011511323.2 3 - 4 1545 140
XM_011513022.2 XP_011511324.2 3 - 4 1545 140
XM_011513023.2 XP_011511325.2 3 - 4 1545 140
XM_017006898.1 XP_016862387.1 3 - 4 1545 140
XM_017006899.1 XP_016862388.1 3 - 4 1545 140
XM_017006900.1 XP_016862389.1 3 - 4 1545 140
XM_017006901.1 XP_016862390.1 3 - 4 1545 140
XM_017006902.1 XP_016862391.1 3 - 4 1545 140
XM_017006903.1 XP_016862392.1 3 - 4 1545 140
XM_017006904.1 XP_016862393.1 3 - 4 1545 140
XM_017006905.1 XP_016862394.1 5 - 6 1001 140
XM_017006906.1 XP_016862395.1 5 - 6 782 140
XM_017006907.1 XP_016862396.1 5 - 6 547 140
XM_017006908.1 XP_016862397.1 5 - 6 672 140
XM_017006909.1 XP_016862398.1 4 - 5 459 140
XM_017006910.1 XP_016862399.1 3 - 4 572 140
XM_017006911.1 XP_016862400.1 4 - 5 849 140
XM_017006912.1 XP_016862401.1 3 - 4 200 198
XM_017006914.1 XP_016862403.1 3 - 4 407 140
XM_017006915.1 XP_016862404.1 3 - 4 389 140
GenBank mRNA AF159441.1 - 4 - 5 439 140
AK301909.1 - 5 - 6 765 140
BC055415.1 - 3 - 4 386 140
BC069785.1 - 3 - 4 332 140
BC120969.2 - 3 - 4 404 140
BC141969.1 - 3 - 4 404 140
BU852276.1 - 3 - 4 385 140

View Full Product Details