Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MIRLET7BHG
- Entrez Gene ID
-
400931
- Gene Name
- MIRLET7B host gene
- Gene Aliases
-
linc-Ppara
- Chromosome Location
-
Chr.22: 46085997 - 46113928 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110479.1 |
|
3 - 4 |
310 |
55 |
GenBank mRNA |
AK130875.1 |
- |
3 - 4 |
304 |
55 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 3619 |
MIR3619 |
|
-7.4kb
|
Sense |
Chr. - 46091044 - 46091126 on Build GRCh38 |
Downstream |
microRNA let-7a-3 |
MIRLET7A3 |
LET7A3, MIRNLET7A3, let-7a-3 |
3.4kb
|
Sense |
Chr. - 46112749 - 46112822 on Build GRCh38 |
View Full Product Details