Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC37-AS1
- Entrez Gene ID
-
100506907
- Gene Name
- CCDC37 antisense RNA 1 (head to head)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 126393032 - 126394798 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103789.1 |
|
1 - 2 |
506 |
61 |
GenBank mRNA |
HY225164.1 |
- |
1 - 2 |
|
61 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA972296.1 |
- |
1 - 2 |
|
|
Chr. 3 - 126393033 - 126394397 [+] on Build GRCh38 |
61 |
GenBankmRNA |
HY191041.1 |
- |
1 - 2 |
|
|
Chr. 3 - 126393033 - 126394409 [+] on Build GRCh38 |
61 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
Kruppel like factor 15 |
KLF15 |
KKLF |
-35.6kb
|
Sense |
Chr. - 126288121 - 126357393 on Build GRCh38 |
Downstream |
cilia and flagella associated protein 100 |
CFAP100 |
CCDC37, MIA1 |
95.0
|
Anti-sense |
Chr. - 126394893 - 126436556 on Build GRCh38 |
View Full Product Details