Product Details

Assay Reference Genome
Location

Chr.7:100403036 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Within Intron 16
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK000919.1 BAA91424.1
AK093038.1 BAC04028.1
AK300745.1
AK302108.1
AL136735.1 CAB66669.1
BC002725.1 AAH02725.1
KU178677.1
KU178678.1
KU178679.1
KU178680.1
KU178681.1
KU178682.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion MUC17 TRIM56 NAT16 ZASP MIR106B GIGYF1 EPHB4 PILRB STAG3L5P LOC105375429 CNPY4 VGF MBLAC1 GAL3ST4 FIS1 MOSPD3 PVRIG AP1S1 C7orf43 C7orf61 PPP1R35 ACHE PILRA MUC3A MOGAT3 ZNHIT1 ZNF3 MIR93 POP7 MCM7 MIR6840 MUC12 ACTL6B NYAP1 AGFG2 MIR4658 GATS TFR2 MIR25 GPC2 MIR4653 TRIP6 EPO STAG3 PCOLCE MEPCE CLDN15 LAMTOR4 ZKSCAN1 AZGP1 ZAN SPDYE3 UFSP1 SRRT FBXO24 PMS2P1 AZGP1P1 PLOD3 TAF6 GNB2 LRCH4 ZCWPW1 SLC12A9 LOC102724094 SAP25 PCOLCE-AS1 SERPINE1 MIR6875 ZSCAN21 COPS6 AP4M1 STAG3L5P-PVRIG2P-PILRB PVRIG2P TSC22D4
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss C7orf61 PPP1R35 PILRA MEPCE PILRB STAG3L5P STAG3L5P-PVRIG2P-PILRB MIR6840 SPDYE3 PVRIG2P NYAP1 TSC22D4 PMS2P1 ZCWPW1

View Full Product Details