Product Details

Assay Reference Genome
Location

Chr.8:31072457 on build GRCh38
Cytoband
8p12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

For this assay, SNP(s) [rs75027493] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Target Gene Details

Entrez Gene ID
7486
Gene Symbol
WRN
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
Location
Chr.8:31033262-31173761 on build GRCh38
Assay Gene Location
Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRN NM_000553.4 NP_000544.2
XM_011544639.2 XP_011542941.1
AF091214.1 AAC63361.1
AY818673.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831278 Chr8:31059468 - 31213390 on Build GRCh38 Gain WRN

View Full Product Details