Product Details

Assay Reference Genome
Location

Chr.16:13938812 on build GRCh38
Cytoband
16p13.12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2072
Gene Symbol
ERCC4
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
Location
Chr.16:13920138-13952348 on build GRCh38
Assay Gene Location
Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ERCC4 NM_005236.2 NP_005227.1
XM_011522424.2 XP_011520726.1
XM_011522427.1 XP_011520729.1
XM_017023043.1 XP_016878532.1
AK289726.1
AK301930.1
U64315.1 AAB07689.1

View Full Product Details