Product Details

Assay Reference Genome
Location

Chr.21:25903676 on build GRCh38
Cytoband
21q21.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
351
Gene Symbol
APP
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Location
Chr.21:25880550-26171128 on build GRCh38
Assay Gene Location
Within Intron 17
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
APP NM_000484.3 NP_000475.1
NM_001136016.3 NP_001129488.1
NM_001136129.2 NP_001129601.1
NM_001136130.2 NP_001129602.1
NM_001136131.2 NP_001129603.1
NM_001204301.1 NP_001191230.1
NM_001204302.1 NP_001191231.1
NM_001204303.1 NP_001191232.1
NM_201413.2 NP_958816.1
NM_201414.2 NP_958817.1
A02759.1 CAA00268.1
AF282245.1 AAQ14327.1
AK294534.1
AK295373.1
AK295621.1
AK296229.1
AK297229.1
AK297412.1
AK298861.1
AK312326.1
BC065529.1 AAH65529.1
M16765.1 AAA51722.1
M18734.1 AAA51726.1
X06989.1 CAA30050.1
Y00264.1 CAA68374.1

View Full Product Details