Product Details

Assay Reference Genome
Location

Chr.X:107635282 on build GRCh38
Cytoband
Xq22.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5631
Gene Symbol
PRPS1
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
Location
Chr.X:107628424-107651026 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRPS1 NM_001204402.1 NP_001191331.1
NM_002764.3 NP_002755.1
AK223169.1 BAD96889.1
AK297968.1
AK307748.1
AK312706.1
AK316467.1
BC001605.1 AAH01605.1
D00860.1 BAA00733.1
DC411129.1
X15331.1 CAA33386.1

View Full Product Details