Product Details
- Assay Reference Genome
Location
Chr.18:51030283 on build GRCh38
- Cytoband
- 18q21.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 4089
- Gene Symbol
- SMAD4
- Gene Name
- SMAD family member 4
- Gene Aliases
- DPC4, JIP, MADH4, MYHRS
- Location
- Chr.18:51030213-51085042 on build GRCh38
- Assay Gene Location
- Within Exon 1
View Full Product Details