Product Details

Assay Reference Genome
Location

Chr.1:202953095 on build GRCh38
Cytoband
1q32.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
51094
Gene Symbol
ADIPOR1
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
Location
Chr.1:202940825-202958572 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ADIPOR1 NM_001290553.1 NP_001277482.1
NM_001290557.1 NP_001277486.1
NM_001290629.1 NP_001277558.1
NM_015999.5 NP_057083.2
XM_017001407.1 XP_016856896.1
AF125179.1 AAQ13552.1
AF125180.1 AAQ13553.1
AF151803.1 AAD34040.1
AK001484.1
AK058114.1
AK222503.1 BAD96223.1
AY424279.1 AAR08367.1
BC001594.2 AAH01594.1
BC010743.1 AAH10743.1
BI597715.1
BX461217.2
DA523648.1

View Full Product Details