Gene Transcripts

Gene Symbol
Cd93
Entrez Gene ID
17064
Gene Name
CD93 antigen
Gene Aliases
6030404G09Rik, AA145088, AA4.1, AW555904, C1qr1, C1qrp, Ly68
Chromosome Location
Chr. 2: 148436651 - 148443535 on Build GRCm38
UniGene ID
Mm.681
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010740.3 NP_034870.1 1 1582
GenBank mRNA AF081789.1 - 1 1591
AK028485.1 - 1 1611
AK079060.1 - 1 1583
AK155060.1 - 1 1484

View Full Product Details