Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039543.2 | NP_001034632.1 | 2 | 232 | |
NM_010801.3 | NP_034931.2 | 2 | 232 | ||
GenBank mRNA | AF009515.1 | - | 2 | 173 | |
AF100171.1 | - | 2 | 173 | ||
AK007292.1 | - | 2 | 232 | ||
AK133448.1 | - | 2 | 233 | ||
BC027526.1 | - | 2 | 181 | 1495721 |