Gene Transcripts

Gene Symbol
Phf3
Entrez Gene ID
213109
Gene Name
PHD finger protein 3
Gene Aliases
2310061N19Rik, mKIAA0244
Chromosome Location
Chr. 1: 30802342 - 30873923 on Build GRCm38
UniGene ID
Mm.194486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081080.1 NP_001074549.1 3 1238
GenBank mRNA BC011226.1 - 3 1233
BC032205.1 - 3 1252
BC137913.1 - 3 1244 8860932

View Full Product Details