Gene Transcripts

Gene Symbol
Rtp4
Entrez Gene ID
67775
Gene Name
receptor transporter protein 4
Gene Aliases
5830458K16Rik, Ifrg28
Chromosome Location
Chr. 16: 23601379 - 23614222 on Build GRCm38
UniGene ID
Mm.475107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023386.5 NP_075875.3 2 579
GenBank mRNA AJ251364.1 - 2 574
AK007477.1 - 2 557
AK018021.1 - 2 572
AY562228.1 - 2 509

View Full Product Details