Gene Transcripts
- Gene Symbol
- Nop9
- Entrez Gene ID
- 67842
- Gene Name
- NOP9 nucleolar protein
- Gene Aliases
- 2610027L16Rik
- Chromosome Location
- Chr. 14: 55745693 - 55755634 on Build GRCm38
- UniGene ID
- Mm.180338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026403.3 |
NP_080679.3
|
1, 2 |
334 |
|
GenBank mRNA |
AK011570.1 |
- |
1, 2 |
324 |
|
AK015585.1 |
- |
1, 2 |
84 |
|
AK032873.1 |
- |
1, 2 |
318 |
|
AK037831.1 |
- |
1, 2 |
322 |
|
AK050912.1 |
- |
1, 2 |
324 |
|
AK083861.1 |
- |
1, 2 |
203 |
|
AK139689.1 |
- |
1, 2 |
284 |
|
AK146020.1 |
- |
1, 2 |
289 |
|
AK146687.1 |
- |
1, 2 |
305 |
|
AK152732.1 |
- |
1, 2 |
308 |
|
AK154387.1 |
- |
1, 2 |
326 |
|
AK159798.1 |
- |
1, 2 |
308 |
|
AK185037.1 |
- |
|
20 |
|
AK219900.1 |
- |
|
20 |
|
BC018523.1 |
- |
1, 2 |
314 |
3499986 |
CJ098414.1 |
- |
1, 2 |
335 |
|
View Full Product Details