Gene Transcripts

Gene Symbol
Chmp2b
Entrez Gene ID
68942
Gene Name
charged multivesicular body protein 2B
Gene Aliases
1190006E07Rik
Chromosome Location
Chr. 16: 65539133 - 65562697 on Build GRCm38
UniGene ID
Mm.432944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026879.2 NP_081155.1 4 547
GenBank mRNA AK004506.1 - 1 20
AK084111.1 - 4 549
BC042626.1 - 3 303
BC055809.1 - 4 576 6414077

View Full Product Details