Gene Transcripts

Gene Symbol
Dsg1c
Entrez Gene ID
211924
Gene Name
desmoglein 1 gamma
Gene Aliases
9330156N18, Dsg1gamma, Dsg6, dsg1g
Chromosome Location
Chr. 18: 20247340 - 20283923 on Build GRCm38
UniGene ID
Mm.391118
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181680.1 NP_859008.1 11 1622
GenBank mRNA AK034104.1 - 1565
AY192159.1 - 11 1622
AY314983.1 - 11 1522

View Full Product Details