Gene Transcripts

Gene Symbol
Gstm3
Entrez Gene ID
14864
Gene Name
glutathione S-transferase, mu 3
Gene Aliases
Fsc2, mGSTM5
Chromosome Location
Chr. 3: 107963696 - 107969174 on Build GRCm38
UniGene ID
Mm.440885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010359.2 NP_034489.1 2 104
GenBank mRNA BC044927.1 - 2 88
J03953.1 - 1 44

View Full Product Details