Gene Transcripts

Gene Symbol
Adgrg6
Entrez Gene ID
215798
Gene Name
adhesion G protein-coupled receptor G6
Gene Aliases
1190004A11Rik, AI449247, AW045736, DREG, Gm222, Gpr126
Chromosome Location
Chr. 10: 14402580 - 14545722 on Build GRCm38
UniGene ID
Mm.440142
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002268.3 NP_001002268.1 3 469
GenBank mRNA AB183545.1 - 3 470

View Full Product Details