Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159392.1 | NP_001152864.1 | 7 | 1043 | |
NM_009395.4 | NP_033421.3 | 7 | 1056 | ||
GenBank mRNA | AK004593.2 | - | 7 | 970 | |
AK034305.1 | - | 7 | 991 | ||
AK147809.1 | - | 7 | 951 | ||
AK168386.1 | - | 7 | 935 | ||
AK168704.1 | - | 7 | 978 | ||
AK168805.1 | - | 7 | 952 | ||
AK172287.1 | - | 4 | 299 | ||
BC003906.1 | - | 7 | 892 | 3484017 |