Gene Transcripts
- Gene Symbol
- Mrgprb1
- Entrez Gene ID
- 233231
- Gene Name
- MAS-related GPR, member B1
- Gene Aliases
- MrgB1
- Chromosome Location
- Chr. 7: 48444113 - 48456342 on Build GRCm38
- UniGene ID
- Mm.297430
- Species
- Mus musculus
- Gene Symbol
- Trpm1
- Entrez Gene ID
- 17364
- Gene Name
- transient receptor potential cation channel, subfamily M, member 1
- Gene Aliases
- 4732499L03Rik, AI606771, Ltrpc1, Mlsn1
- Chromosome Location
- Chr. 7: 64153835 - 64269772 on Build GRCm38
- UniGene ID
- Mm.297430
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK192880.1 |
- |
|
71 |
|
View Full Product Details