Gene Transcripts

Gene Symbol
Loxhd1
Entrez Gene ID
240411
Gene Name
lipoxygenase homology domains 1
Gene Aliases
1700096C21Rik, 4932417K07, sba
Chromosome Location
Chr. 18: 77281949 - 77442932 on Build GRCm38
UniGene ID
Mm.380247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172834.2 NP_766422.2 17 2361
GenBank mRNA AK030047.1 - 14 2196
FJ750876.1 - 17 2361

View Full Product Details