Gene Transcripts

Gene Symbol
Taf1
Entrez Gene ID
270627
Gene Name
TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
AU015687, B430306D02Rik, Ccg-1, Ccg1, KAT4, N-TAF1, TAFII250, Taf2a, p250
Chromosome Location
Chr. X: 101527574 - 101601789 on Build GRCm38
UniGene ID
Mm.261750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290729.1 NP_001277658.1 17 2803
GenBank mRNA AB299229.1 - 17 2730
AF022178.1 - 2 118
AK050691.1 - 2 174
BC047418.1 - 7 916
BC051073.1 - 4 529

View Full Product Details