Gene Transcripts

Gene Symbol
Taar9
Entrez Gene ID
503558
Gene Name
trace amine-associated receptor 9
Gene Aliases
Gm231, taR-9
Chromosome Location
Chr. 10: 24108488 - 24109534 on Build GRCm38
UniGene ID
Mm.300923
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010831.1 NP_001010831.1 1 1003
GenBank mRNA BC139084.1 - 1 1052
BC139085.1 - 1 1051 8862107

View Full Product Details