Gene Transcripts

Gene Symbol
Hspbp1
Entrez Gene ID
66245
Gene Name
HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1
Gene Aliases
1500019G21Rik
Chromosome Location
Chr. 7: 4660515 - 4685121 on Build GRCm38
UniGene ID
Mm.6803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024172.3 NP_077134.1 7 1071
GenBank mRNA AF338351.1 - 6 1009
AK013263.1 - 7 1062
AK088469.1 - 7 1047
AK160128.1 - 7 1067

View Full Product Details