Gene Transcripts

Gene Symbol
Syce1l
Entrez Gene ID
668110
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
4930481F22Rik, MMRP, PSESS, mmrp2
Chromosome Location
Chr. 8: 113643213 - 113655533 on Build GRCm38
UniGene ID
Mm.390078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048145.1 NP_001041610.1 4, 5 283
NM_001286494.1 NP_001273423.1 3, 4 220
NM_029139.1 NP_083415.1 4, 5 283
GenBank mRNA AK015606.1 - 4, 5 279
AY899935.1 - 4, 5 283
AY899936.1 - 4, 5 283
BB616021.1 - 4, 5 263
BC100478.1 - 3, 4 201
BC141326.1 - 4, 5 283 9055938
BC141347.1 - 4, 5 283 9055959

View Full Product Details