Gene Transcripts

Gene Symbol
Syce1l
Entrez Gene ID
668110
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
4930481F22Rik, MMRP, PSESS, mmrp2
Chromosome Location
Chr. 8: 113643213 - 113655533 on Build GRCm38
UniGene ID
Mm.390078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048145.1 NP_001041610.1 9, 10 618
NM_001286494.1 NP_001273423.1 8, 9 555
NM_029139.1 NP_083415.1 10, 11 682
GenBank mRNA AK015606.1 - 10, 11 678
AY899935.1 - 9, 10 618
AY899936.1 - 10, 11 682
BC100478.1 - 8, 9 536
BC141326.1 - 9, 10 618 9055938
BC141347.1 - 9, 10 618 9055959

View Full Product Details