Gene Transcripts

Gene Symbol
Fbxo24
Entrez Gene ID
71176
Gene Name
F-box protein 24
Gene Aliases
4933422D21Rik, Fbx24
Chromosome Location
Chr. 5: 137612505 - 137625584 on Build GRCm38
UniGene ID
Mm.474695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027708.1 NP_081984.1 2 209
GenBank mRNA AK016870.1 - 2 210

View Full Product Details