Gene Transcripts

Gene Symbol
Ulk3
Entrez Gene ID
71742
Gene Name
unc-51-like kinase 3
Gene Aliases
1200015E14Rik
Chromosome Location
Chr. 9: 57589137 - 57596233 on Build GRCm38
UniGene ID
Mm.155877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027895.1 NP_082171.1 3 432
GenBank mRNA AK004783.1 - 3 416
AK032030.1 - 1 53
AK154642.1 - 3 345
BC037093.1 - 3 453
BC151153.1 - 3 358 9088047

View Full Product Details