Gene Transcripts

Gene Symbol
Tmem87b
Entrez Gene ID
72477
Gene Name
transmembrane protein 87B
Gene Aliases
2610301K12Rik, 2810431I02Rik, AU014804
Chromosome Location
Chr. 2: 128818072 - 128854261 on Build GRCm38
UniGene ID
Mm.259688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028248.2 NP_082524.2 4 376
GenBank mRNA AK011959.1 - 4 297
AK050203.1 - 4 368
AK164504.1 - 4 358
AK166991.1 - 4 356
AK171811.1 - 5 292
AK172611.1 - 5 292
BC031488.1 - 4 332
CA479832.1 - 4 598

View Full Product Details