Product Details

SNP ID
rs13481248
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:116126213 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGCATAGAAAGTGTGCAGCGGCTG[C/T]GTGCGTGGCTCCAGGCTGTAGAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Mrpl38 PubMed Links

Gene Details

Gene
Mrpl38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
Trim65
Gene Name
tripartite motif-containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178802.4 1545 Silent Mutation NP_848917.2

View Full Product Details